Assay Details
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105998.1 | NP_001099468.1 | 6 - 7 | 944 | 83 | |
XM_006250715.2 | XP_006250777.1 | 5 - 6 | 1044 | 83 | ||
GenBank mRNA | FQ223715.1 | - | 5 - 6 | 1037 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P123316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150950 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123316 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123316 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P123316 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P109887 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P268325 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10771590 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771591 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771587 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771586 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771589 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771585 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771588 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10771584 | - | - |
Rat Genome 230 2.0 Array | 1392937_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392937_at | - | - |
Rat Genome 230 2.0 Array | 1388370_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388370_at | - | - |
Affymetrix HT Rat Focus Array | 1388370_at | - | - |
Affymetrix HT Rat Focus Array | 1388370_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392937_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392937_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400001842.rn.1 | TC1400000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001843.rn.1 | TC1400000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001841.rn.1 | TC1400000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001847.rn.1 | TC1400000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001840.rn.1 | TC1400000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001846.rn.1 | TC1400000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001845.rn.1 | TC1400000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001844.rn.1 | TC1400000237.rn.1 | RTA TC |
Rat Expression Set 230 | 1388370_at | - | - |
Rat Expression Set 230 | 1388370_PM_at | - | - |
Rat Expression Set 230 | 1392937_PM_at | - | - |
Rat Expression Set 230 | 1392937_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771584 | - | - |
Rat Genome U34 Set | rc_AA943280_f_at | - | - |
Rat Genome U34 Set | rc_AI176278_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21069597 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355251 | - | - |
