Assay Details
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105998.1 | NP_001099468.1 | 1 - 2 | 118 | 141 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001105998.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
