Assay Details
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105998.1 | NP_001099468.1 | 2 - 3 | 444 | 112 | |
XM_006250715.2 | XP_006250777.1 | 1 - 2 | 544 | 112 | ||
GenBank mRNA | FQ223715.1 | - | 1 - 2 | 537 | 112 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001105998.1, XM_006250715.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
