Assay Details
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013131.1 | NP_001013149.1 | 1 - 2 | 63 | 59 | |
GenBank mRNA | BC091207.1 | - | 1 - 2 | 63 | 7381272 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
