Assay Details
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013131.1 | NP_001013149.1 | 5 - 6 | 653 | 85 | |
GenBank mRNA | BC091207.1 | - | 5 - 6 | 653 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
