Assay Details
Gene Symbol: Rab2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037645.1 | NP_001032734.1 | 1 - 2 | 141 | 92 | |
XM_006251883.2 | XP_006251945.1 | 1 - 2 | 211 | 92 | ||
GenBank mRNA | BC107443.1 | - | 1 - 2 | 141 | 7307807 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC107443.1, NM_001037645.1, XM_006251883.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
