Assay Details
Gene Symbol: Gnpnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251782.2 | XP_006251844.1 | 1 - 2 | 94 | 105 | |
XM_006251784.2 | XP_006251846.1 | 1 - 2 | 90 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006251782.2, XM_006251784.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
