Assay Details
Gene Symbol: Cdkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251791.2 | XP_006251853.1 | 5 - 6 | 503 | 72 | |
XM_006251792.2 | XP_006251854.1 | 5 - 6 | 503 | 72 | ||
GenBank mRNA | BC167026.1 | - | 4 - 5 | 417 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
