Assay Details
Gene Symbol: Peli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107259.1 | NP_001100729.1 | 5 - 6 | 953 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P043285 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P043289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46418 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043289 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043289 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P043289 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P610733 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1020477 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18806 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99187 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64262 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10779727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779729 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779730 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10779726 | - | - |
Rat Genome 230 2.0 Array | 1390942_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384706_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390942_at | - | - |
Rat Genome 230 2.0 Array | 1384706_at | - | - |
Affymetrix HT Rat Focus Array | 1384706_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390942_at | - | - |
Affymetrix HT Rat Focus Array | 1390942_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384706_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500001832.rn.1 | TC1500000255.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500001835.rn.1 | TC1500000255.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500001831.rn.1 | TC1500000255.rn.1 | RTA TC |
Rat Expression Set 230 | 1384706_PM_at | - | - |
Rat Expression Set 230 | 1384706_at | - | - |
Rat Expression Set 230 | 1390942_at | - | - |
Rat Expression Set 230 | 1390942_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779726 | - | - |
Rat Genome U34 Set | rc_AI102884_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21191537 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357893 | - | - |
