Assay Details
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024794.1 | NP_001019965.1 | 5 - 6 | 1186 | 58 | |
XM_008770555.1 | XP_008768777.1 | 5 - 6 | 1559 | 58 | ||
GenBank mRNA | BC097400.1 | - | 5 - 6 | 1186 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
