Assay Details
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277600.1 | NP_001264529.1 | 1 - 2 | 152 | 105 | |
NM_001277601.1 | NP_001264530.1 | 1 - 2 | 152 | 105 | ||
XM_006251866.1 | XP_006251928.1 | 1 - 2 | 139 | 105 | ||
XM_008770510.1 | XP_008768732.1 | 1 - 2 | 147 | 105 | ||
XM_008770511.1 | XP_008768733.1 | 1 - 2 | 147 | 105 | ||
XM_008770512.1 | XP_008768734.1 | 1 - 2 | 139 | 105 | ||
GenBank mRNA | CB582273.1 | - | 1 - 2 | 102 | 105 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271241.1 | NP_001258170.1 | 1 - 2 | 117 | 105 | |
XM_003751394.2 | XP_003751442.1 | 1 - 2 | 103 | 105 | ||
XM_003751395.2 | XP_003751443.1 | 1 - 2 | 103 | 105 | ||
XM_006251452.2 | XP_006251514.1 | 1 - 2 | 103 | 105 | ||
XM_006251458.2 | XP_006251520.1 | 1 - 2 | 103 | 105 | ||
XM_008770243.1 | XP_008768465.1 | 1 - 2 | 103 | 105 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001271241.1,NM_001277600.1,NM_001277601.1,XM_003751394.2,XM_003751395.2,XM_006251452.2,XM_006251458.2,XM_006251866.1,XM_008770243.1,XM_008770510.1,XM_008770511.1,XM_008770512.1 transcripts.
This assay is in the 5'UTR region of the NM_001271241.1,NM_001277600.1,NM_001277601.1,XM_003751394.2,XM_003751395.2,XM_006251452.2,XM_006251458.2,XM_006251866.1,XM_008770243.1,XM_008770510.1,XM_008770511.1,XM_008770512.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
