Assay Details
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277600.1 | NP_001264529.1 | 5 - 6 | 614 | 86 | |
XM_006251858.2 | XP_006251920.1 | 4 - 5 | 1082 | 86 | ||
XM_006251859.2 | XP_006251921.1 | 4 - 5 | 575 | 86 | ||
XM_008770508.1 | XP_008768730.1 | 5 - 6 | 1109 | 86 | ||
XM_008770509.1 | XP_008768731.1 | 5 - 6 | 2177 | 86 | ||
XM_008770510.1 | XP_008768732.1 | 5 - 6 | 609 | 86 | ||
XM_008770511.1 | XP_008768733.1 | 5 - 6 | 609 | 86 | ||
GenBank mRNA | CB582273.1 | - | 5 - 6 | 564 | 86 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751395.2 | XP_003751443.1 | 5 - 6 | 565 | 86 | |
XM_006251449.2 | XP_006251511.1 | 5 - 6 | 2176 | 86 | ||
XM_006251450.2 | XP_006251512.1 | 4 - 5 | 1080 | 86 | ||
XM_006251451.1 | XP_006251513.1 | 4 - 5 | 539 | 86 | ||
XM_006251452.2 | XP_006251514.1 | 5 - 6 | 565 | 86 | ||
XM_008770242.1 | XP_008768464.1 | 5 - 6 | 1073 | 86 | ||
XM_008770243.1 | XP_008768465.1 | 5 - 6 | 565 | 86 | ||
GenBank mRNA | FQ215781.1 | - | 3 - 4 | 264 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
