Assay Details
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277600.1 | NP_001264529.1 | 6 - 7 | 782 | 134 | |
NM_001277601.1 | NP_001264530.1 | 6 - 7 | 743 | 134 | ||
NM_001277602.1 | NP_001264531.1 | 5 - 6 | 717 | 134 | ||
XM_006251858.2 | XP_006251920.1 | 5 - 6 | 1250 | 134 | ||
XM_006251859.2 | XP_006251921.1 | 5 - 6 | 743 | 134 | ||
XM_006251861.2 | XP_006251923.1 | 5 - 6 | 1210 | 134 | ||
XM_006251862.2 | XP_006251924.1 | 7 - 8 | 894 | 134 | ||
XM_006251863.2 | XP_006251925.1 | 5 - 6 | 704 | 134 | ||
XM_006251864.2 | XP_006251926.1 | 7 - 8 | 799 | 134 | ||
XM_006251865.2 | XP_006251927.1 | 5 - 6 | 1210 | 134 | ||
XM_006251866.1 | XP_006251928.1 | 6 - 7 | 730 | 134 | ||
XM_008770508.1 | XP_008768730.1 | 6 - 7 | 1277 | 134 | ||
XM_008770509.1 | XP_008768731.1 | 6 - 7 | 2345 | 134 | ||
XM_008770510.1 | XP_008768732.1 | 6 - 7 | 777 | 134 | ||
XM_008770511.1 | XP_008768733.1 | 6 - 7 | 777 | 134 | ||
XM_008770512.1 | XP_008768734.1 | 6 - 7 | 730 | 134 | ||
GenBank mRNA | FQ214011.1 | - | 4 - 5 | 351 | 134 | |
FQ227266.1 | - | 4 - 5 | 393 | 134 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271241.1 | NP_001258170.1 | 6 - 7 | 708 | 134 | |
XM_003751394.2 | XP_003751442.1 | 6 - 7 | 694 | 134 | ||
XM_003751395.2 | XP_003751443.1 | 6 - 7 | 733 | 134 | ||
XM_006251449.2 | XP_006251511.1 | 6 - 7 | 2344 | 134 | ||
XM_006251450.2 | XP_006251512.1 | 5 - 6 | 1248 | 134 | ||
XM_006251451.1 | XP_006251513.1 | 5 - 6 | 707 | 134 | ||
XM_006251452.2 | XP_006251514.1 | 6 - 7 | 733 | 134 | ||
XM_006251453.1 | XP_006251515.1 | 7 - 8 | 858 | 134 | ||
XM_006251454.1 | XP_006251516.1 | 7 - 8 | 763 | 134 | ||
XM_006251455.2 | XP_006251517.1 | 5 - 6 | 1208 | 134 | ||
XM_006251456.1 | XP_006251518.1 | 5 - 6 | 668 | 134 | ||
XM_006251457.1 | XP_006251519.1 | 5 - 6 | 668 | 134 | ||
XM_006251458.2 | XP_006251520.1 | 6 - 7 | 694 | 134 | ||
XM_008770242.1 | XP_008768464.1 | 6 - 7 | 1241 | 134 | ||
XM_008770243.1 | XP_008768465.1 | 6 - 7 | 733 | 134 | ||
GenBank mRNA | AY724499.1 | - | 5 - 6 | 1136 | 134 | |
FQ214198.1 | - | 4 - 5 | 393 | 134 | ||
FQ215781.1 | - | 4 - 5 | 432 | 134 | ||
FQ227893.1 | - | 4 - 5 | 393 | 134 | ||
FQ228082.1 | - | 4 - 5 | 432 | 134 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
