Assay Details
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100509.1 | NP_001093979.1 | 7 - 8 | 581 | 96 | |
GenBank mRNA | BC107950.1 | - | 7 - 8 | 502 | 96 | |
DY570284.1 | - | 7 - 8 | 581 | 96 | ||
FQ213949.1 | - | 7 - 8 | 529 | 96 | ||
FQ214043.1 | - | 7 - 8 | 534 | 96 | ||
FQ227742.1 | - | 7 - 8 | 538 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
