Assay Details
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107254.1 | NP_001100724.1 | 1 - 2 | 301 | 117 | |
XM_008770515.1 | XP_008768737.1 | 1 - 2 | 1139 | 117 | ||
XM_008770516.1 | XP_008768738.1 | 1 - 2 | 1138 | 117 | ||
GenBank mRNA | BC168894.1 | - | 1 - 2 | 283 | 7453870 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC168894.1, NM_001107254.1, XM_008770515.1, XM_008770516.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
