Assay Details
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107254.1 | NP_001100724.1 | 7 - 8 | 1643 | 113 | |
XM_008770515.1 | XP_008768737.1 | 7 - 8 | 2520 | 113 | ||
XM_008770516.1 | XP_008768738.1 | 6 - 7 | 2255 | 113 | ||
GenBank mRNA | BC168894.1 | - | 7 - 8 | 1664 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
