Assay Details
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107254.1 | NP_001100724.1 | 9 - 10 | 1964 | 78 | |
XM_008770515.1 | XP_008768737.1 | 9 - 10 | 2841 | 78 | ||
XM_008770516.1 | XP_008768738.1 | 8 - 9 | 2576 | 78 | ||
GenBank mRNA | BC168894.1 | - | 9 - 10 | 1985 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
