Assay Details
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770516.1 | XP_008768738.1 | 2 - 3 | 1659 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
