Assay Details
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173324.2 | NP_775446.2 | 3 - 4 | 370 | 155 | |
GenBank mRNA | AF267197.1 | - | 2 - 3 | 322 | 155 | |
BC081696.1 | - | 3 - 4 | 370 | 155 | ||
FQ212245.1 | - | 3 - 4 | 378 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
