Assay Details
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173324.2 | NP_775446.2 | 8 - 9 | 1855 | 192 | |
GenBank mRNA | BC081696.1 | - | 8 - 9 | 1855 | 7112549 | 192 |
FQ210274.1 | - | 8 - 9 | 1863 | 192 | ||
FQ212245.1 | - | 8 - 9 | 1863 | 192 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
