Assay Details
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109082.1 | NP_001102552.1 | 1 - 2 | 83 | 168 | |
XM_006250463.2 | XP_006250525.1 | 1 - 2 | 98 | 168 | ||
XM_006250465.2 | XP_006250527.1 | 1 - 2 | 98 | 168 | ||
XM_008769868.1 | XP_008768090.1 | 1 - 2 | 98 | 168 | ||
XM_008769870.1 | XP_008768092.1 | 1 - 2 | 98 | 168 | ||
XM_008769871.1 | XP_008768093.1 | 1 - 2 | 98 | 168 | ||
XM_008769872.1 | XP_008768094.1 | 1 - 2 | 98 | 168 | ||
GenBank mRNA | AY724511.1 | - | 1 - 2 | 43 | 168 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109082.1, XM_006250463.2, XM_006250465.2, XM_008769868.1, XM_008769870.1, XM_008769871.1, XM_008769872.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
