Assay Details
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109082.1 | NP_001102552.1 | 3 - 4 | 581 | 83 | |
XM_006250463.2 | XP_006250525.1 | 3 - 4 | 596 | 83 | ||
XM_006250465.2 | XP_006250527.1 | 3 - 4 | 596 | 83 | ||
XM_008769868.1 | XP_008768090.1 | 3 - 4 | 596 | 83 | ||
XM_008769869.1 | XP_008768091.1 | 3 - 4 | 919 | 83 | ||
XM_008769870.1 | XP_008768092.1 | 3 - 4 | 596 | 83 | ||
XM_008769871.1 | XP_008768093.1 | 3 - 4 | 596 | 83 | ||
XM_008769872.1 | XP_008768094.1 | 3 - 4 | 596 | 83 | ||
GenBank mRNA | AY724511.1 | - | 3 - 4 | 541 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
