Assay Details
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250463.2 | XP_006250525.1 | 5 - 6 | 725 | 91 | |
XM_006250465.2 | XP_006250527.1 | 5 - 6 | 725 | 91 | ||
XM_008769868.1 | XP_008768090.1 | 5 - 6 | 725 | 91 | ||
XM_008769869.1 | XP_008768091.1 | 5 - 6 | 1048 | 91 | ||
XM_008769870.1 | XP_008768092.1 | 5 - 6 | 725 | 91 | ||
XM_008769871.1 | XP_008768093.1 | 5 - 6 | 725 | 91 | ||
XM_008769872.1 | XP_008768094.1 | 5 - 6 | 725 | 91 | ||
GenBank mRNA | AY724511.1 | - | 5 - 6 | 670 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
