Assay Details
Gene Symbol: Ephx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012844.3 | NP_036976.2 | 1 - 2 | 165 | 76 | |
GenBank mRNA | M26125.1 | - | 141 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M26125.1, NM_012844.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
