Assay Details
Gene Symbol: Camk1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182842.1 | NP_878262.1 | 3 - 4 | 306 | 112 | |
XM_006250470.2 | XP_006250532.1 | 3 - 4 | 423 | 112 | ||
GenBank mRNA | AB101231.1 | - | 3 - 4 | 306 | 112 | |
AB101232.1 | - | 3 - 4 | 306 | 112 | ||
D86557.1 | - | 3 - 4 | 312 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
