Assay Details
Gene Symbol: Arhgap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109077.1 | NP_001102547.1 | 1 - 2 | 171 | 84 | |
GenBank mRNA | FQ211641.1 | - | 179 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
