Assay Details
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031555.1 | NP_113743.1 | 4 - 5 | 617 | 80 | |
XM_006235117.2 | XP_006235179.1 | 4 - 5 | 278 | 80 | ||
GenBank mRNA | AB003104.1 | - | 4 - 5 | 618 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
