Assay Details
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031555.1 | NP_113743.1 | 5 - 6 | 729 | 76 | |
XM_006235117.2 | XP_006235179.1 | 5 - 6 | 390 | 76 | ||
GenBank mRNA | AB003104.1 | - | 5 - 6 | 730 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
