Assay Details
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031555.1 | NP_113743.1 | 6 - 7 | 948 | 116 | |
XM_006235117.2 | XP_006235179.1 | 6 - 7 | 609 | 116 | ||
GenBank mRNA | AB003104.1 | - | 6 - 7 | 949 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
