Assay Details
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031555.1 | NP_113743.1 | 7 - 8 | 1038 | 70 | |
XM_006235117.2 | XP_006235179.1 | 7 - 8 | 699 | 70 | ||
GenBank mRNA | AB003104.1 | - | 7 - 8 | 1039 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P285694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114670 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P285694 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P285694 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P285694 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P285694 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58186 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46764 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39935 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10850357 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850355 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850361 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850360 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850356 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850359 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850354 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850358 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850353 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10850352 | - | - |
Rat Genome 230 2.0 Array | 1388144_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388144_at | - | - |
Affymetrix HT Rat Focus Array | 1388144_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388144_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300033332.rn.1 | TC0300004315.rn.1 | RTA TC |
Rat Expression Set 230 | 1388144_at | - | - |
Rat Expression Set 230 | 1388144_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850352 | - | - |
Rat Genome U34 Set | AB003104_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21068781 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349456 | - | - |
