Assay Details
Gene Symbol: Pax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107787.1 | NP_001101257.1 | 3 - 4 | 1383 | 93 | |
XM_006235152.2 | XP_006235214.2 | 4 - 5 | 2689 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
