Assay Details
Gene Symbol: Polr3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107784.1 | NP_001101254.1 | 3 - 4 | 457 | 140 | |
XM_008762269.1 | XP_008760491.1 | 3 - 4 | 488 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
