Assay Details
Gene Symbol: Polr3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107784.1 | NP_001101254.1 | 8 - 9 | 1082 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P370537 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P807990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102717 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P370537 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P807990 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P807990 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P370537 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P807990 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P370537 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P370537 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90731 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20472 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10840391 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840387 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840389 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840386 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840385 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840388 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840390 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840392 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840393 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10840384 | - | - |
Rat Genome 230 2.0 Array | 1391496_at | - | - |
Rat Genome 230 2.0 Array | 1386008_at | - | - |
Rat Genome 230 2.0 Array | 1386008_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391496_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391496_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391496_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300013505.rn.1 | TC0300001885.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300013506.rn.1 | TC0300001885.rn.1 | RTA TC |
Rat Expression Set 230 | 1386008_at | - | - |
Rat Expression Set 230 | 1386008_PM_at | - | - |
Rat Expression Set 230 | 1391496_PM_at | - | - |
Rat Expression Set 230 | 1391496_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840384 | - | - |
Rat Genome U34 Set | rc_AA819635_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22071701 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360614 | - | - |
