Assay Details
Gene Symbol: Slco4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133608.2 | NP_598292.2 | 8 - 9 | 1814 | 104 | |
GenBank mRNA | AF239262.1 | - | 7 - 8 | 1648 | 104 | |
BC097285.1 | - | 8 - 9 | 1814 | 104 | ||
BC097286.1 | - | 8 - 9 | 1756 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
