Assay Details
Gene Symbol: Dnajc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762544.1 | XP_008760766.1 | 1 - 2 | 155 | 66 | |
GenBank mRNA | FQ223796.1 | - | 1 - 2 | 147 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008762544.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
