Assay Details
Gene Symbol: Dnajc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762545.1 | XP_008760767.1 | 2 - 3 | 207 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008762545.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
