Assay Details
Gene Symbol: Dnajc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024161.2 | NP_077075.1 | 1 - 2 | 104 | 92 | |
GenBank mRNA | AF323955.1 | - | 1 - 2 | 104 | 92 | |
U39320.1 | - | 1 - 2 | 104 | 92 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF323955.1, NM_024161.2, U39320.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
