Assay Details
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762541.1 | XP_008760763.1 | 1 - 2 | 65 | 57 | |
XM_008762542.1 | XP_008760764.1 | 1 - 2 | 44 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008762541.1, XM_008762542.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
