Assay Details
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235801.2 | XP_006235863.1 | 1 - 2 | 2977 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
