Assay Details
Gene Symbol: Psma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008217.1 | NP_001008218.1 | 2 - 3 | 290 | 94 | |
GenBank mRNA | BC127466.1 | - | 2 - 3 | 310 | 94 | |
CF110813.1 | - | 2 - 3 | 244 | 94 | ||
FQ226344.1 | - | 2 - 3 | 297 | 94 | ||
FQ231713.1 | - | 2 - 3 | 297 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
