Assay Details
Gene Symbol: Psma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008217.1 | NP_001008218.1 | 5 - 6 | 661 | 138 | |
GenBank mRNA | BC127466.1 | - | 4 - 5 | 1079 | 138 | |
CF110813.1 | - | 5 - 6 | 615 | 138 | ||
D30804.1 | - | 5 - 6 | 653 | 138 | ||
FQ226344.1 | - | 5 - 6 | 668 | 138 | ||
FQ231713.1 | - | 5 - 6 | 668 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
