Assay Details
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145090.3 | NP_659558.1 | 6 - 7 | 724 | 101 | |
XM_006235731.2 | XP_006235793.1 | 6 - 7 | 824 | 101 | ||
XM_006235732.1 | XP_006235794.1 | 6 - 7 | 765 | 101 | ||
XM_006235733.2 | XP_006235795.1 | 5 - 6 | 1033 | 101 | ||
XM_006235734.2 | XP_006235796.1 | 5 - 6 | 1032 | 101 | ||
GenBank mRNA | BC070895.1 | - | 6 - 7 | 705 | 7099840 | 101 |
DQ167810.1 | - | 4 - 5 | 441 | 101 | ||
DQ167811.1 | - | 4 - 5 | 441 | 101 | ||
FQ222295.1 | - | 6 - 7 | 747 | 101 | ||
FQ232032.1 | - | 6 - 7 | 745 | 101 | ||
FQ233265.1 | - | 6 - 7 | 745 | 101 | ||
U35776.1 | - | 6 - 7 | 724 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
