Assay Details
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145090.3 | NP_659558.1 | 9 - 10 | 963 | 105 | |
XM_006235734.2 | XP_006235796.1 | 8 - 9 | 1271 | 105 | ||
GenBank mRNA | BC070895.1 | - | 9 - 10 | 944 | 7099840 | 105 |
CV120035.1 | - | 1 - 2 | 54 | 105 | ||
FQ222295.1 | - | 9 - 10 | 986 | 105 | ||
U35776.1 | - | 9 - 10 | 963 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
