Assay Details
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031708.1 | NP_113896.1 | 1 - 2 | 61 | 108 | |
XM_006235822.1 | XP_006235884.1 | 1 - 2 | 182 | 108 | ||
GenBank mRNA | AB032742.1 | - | 1 - 2 | 61 | 108 | |
BC061773.1 | - | 1 - 2 | 77 | 5621873 | 108 | |
FQ218007.1 | - | 1 - 2 | 79 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB032742.1, BC061773.1, NM_031708.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
