Assay Details
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031708.1 | NP_113896.1 | 5 - 6 | 611 | 104 | |
XM_006235822.1 | XP_006235884.1 | 5 - 6 | 732 | 104 | ||
GenBank mRNA | AB032742.1 | - | 5 - 6 | 611 | 104 | |
BC061773.1 | - | 5 - 6 | 627 | 5621873 | 104 | |
FQ218007.1 | - | 5 - 6 | 629 | 104 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
