Assay Details
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108254.1 | NP_001101724.1 | 5 - 6 | 717 | 72 | |
XM_008773171.1 | XP_008771393.1 | 5 - 6 | 757 | 72 | ||
XM_008773172.1 | XP_008771394.1 | 5 - 6 | 757 | 72 | ||
GenBank mRNA | BC081944.1 | - | 5 - 6 | 686 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
