Assay Details
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108254.1 | NP_001101724.1 | 8 - 9 | 1299 | 107 | |
XM_008773171.1 | XP_008771393.1 | 8 - 9 | 1339 | 107 | ||
XM_008773172.1 | XP_008771394.1 | 8 - 9 | 1339 | 107 | ||
GenBank mRNA | BC081944.1 | - | 8 - 9 | 1268 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P19435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80683 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P19435 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P19435 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P19435 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19435 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27127 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7383 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38263 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10933358 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933356 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933357 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933352 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933361 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933354 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933360 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933353 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933359 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933355 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933351 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933350 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10933349 | - | - |
Rat Genome 230 2.0 Array | 1393335_at | - | - |
Rat Genome 230 2.0 Array | 1393335_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393335_at | - | - |
Affymetrix HT Rat Focus Array | 1393335_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00002177.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002183.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002181.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002178.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002189.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002188.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002173.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002186.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002176.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002179.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002182.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002187.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002174.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002175.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002190.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002180.rn.1 | TC0X00000291.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00002172.rn.1 | TC0X00000291.rn.1 | RTA TC |
Rat Expression Set 230 | 1393335_PM_at | - | - |
Rat Expression Set 230 | 1393335_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933349 | - | - |
Rat Genome U34 Set | rc_AI070850_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22088596 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361440 | - | - |
