Assay Details
Gene Symbol: Clcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022198.2 | NP_071534.2 | 7 - 8 | 1010 | 150 | |
XM_006256828.2 | XP_006256890.1 | 7 - 8 | 1313 | 150 | ||
GenBank mRNA | BC092209.1 | - | 7 - 8 | 1010 | 7315063 | 150 |
FQ210965.1 | - | 7 - 8 | 1049 | 150 | ||
Z36944.1 | - | 5 - 6 | 718 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
