Assay Details
Gene Symbol: Pik3r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022213.1 | NP_071549.1 | 1 - 2 | 104 | 89 | |
XM_006238714.2 | XP_006238776.1 | 2 - 3 | 125 | 89 | ||
XM_008763979.1 | XP_008762201.1 | 2 - 3 | 327 | 89 | ||
GenBank mRNA | D64047.1 | - | 1 - 2 | 104 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
