Assay Details
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133609.2 | NP_598293.2 | 11 - 12 | 1413 | 151 | |
XM_006238630.2 | XP_006238692.1 | 11 - 12 | 1384 | 151 | ||
GenBank mRNA | BC072507.1 | - | 11 - 12 | 1413 | 7099973 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
translation
translational initiation
regulation of translation
response to heat
response to glucose
oligodendrocyte development
negative regulation of translational initiation in response to stress
response to peptide hormone
positive regulation of GTPase activity
translation initiation factor activity
guanyl-nucleotide exchange factor activity
translation factor activity, RNA binding
nucleotidyltransferase activity
translation
translational initiation
regulation of translation
response to heat
response to glucose
oligodendrocyte development
negative regulation of translational initiation in response to stress
response to peptide hormone
positive regulation of GTPase activity
translation initiation factor activity
guanyl-nucleotide exchange factor activity
translation factor activity, RNA binding
nucleotidyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P151724 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P337088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139603 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151724 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P337088 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P337088 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151724 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P151724 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P337088 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P594008 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P337088 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701603861 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702028309 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61283 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80762 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1228 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44872 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701603861 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702028309 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10871373 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871380 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871371 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871376 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871372 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871379 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871377 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10871369 | - | - |
Rat Genome 230 2.0 Array | 1368967_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368967_at | - | - |
Affymetrix HT Rat Focus Array | 1368967_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368967_at | - | - |
Rat Expression Set 230 | 1368967_PM_at | - | - |
Rat Expression Set 230 | 1368967_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871369 | - | - |
Rat Genome U34 Set | U38253_g_at | - | - |
Rat Genome U34 Set | U38253_at | - | - |
Rat Genome U34 Set | rc_AI639441_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370324 | - | - |
RatRef-12 Expression BeadChip | ILMN_49400 | - | - |
